Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Actual product may vary
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Minimize the risk of cross-contamination with these flex filtered pipette tips that combine the ergonomics of a standard pipette tip with the safety of a filter tip.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
94056970
also known as FS94056970
10 mL
94056520
also known as FS94056520
100 μL
Catalog number 94056970
also known as FS94056970
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
10 mL
Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.
Key Features:
  • Excellent for single and multichannel pipetting
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip Flex pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilized, 10 x 96 rack (10 mL Extended in bag)
Softer Finntip Flex Tip
  • The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination (excluding 10 mL Extended)
Versatile Tip Range
  • Available in volumes: 10, 30, 100, 200, 300, 1000, 1200 μL and 10mL Extended (bottle tip)
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Flex Filter 10 mL Ext., sterile
Volume (Metric)10 mL
FilteredFiltered
SterilitySterile
AutoclavableFinntip pipette tips and racks can be autoclaved for 121 degrees Celsius for 20 minutes
Length (Metric)26.7 cm
Color CodeRed
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Flex
Fit for PurposeCertified Quality
Shelf LifeRecommended shelf life for Finntip pipette tips is five years
Unit SizePK