Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Actual product may vary
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Finntip™ Filtered Pipette Tips
Protect pipettes and the samples from contamination with these filtered pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)
94052200100 μL5.2 cm
9405215020 μL5.2 cm
94052350300 μL5.2 cm
940524101000 μL7.1 cm
9405280010 mL15 cm
940525505 mL14.7 cm
Catalog number 94052200
Price (MYR)
1,159.00
CS
Add to cart
Volume (Metric):
100 μL
Length (Metric):
5.2 cm
Price (MYR)
1,159.00
CS
Add to cart
Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0.2μL-10mL.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 20, 30, 100, 200, 300 and 1000 μL
  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Filter 100 μL, sterile
Volume (Metric)100 μL
FilteredFiltered
SterilitySterile
AutoclavableFinntip pipette tips and racks can be autoclaved for 121 degrees Celsius for 20 minutes
Length (Metric)5.2 cm
FormatRack
Color CodeYellow
DisposableSingle-use
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Filtered
Fit for PurposeSterile & PCR Ready
Shelf LifeRecommended shelf life for Finntip pipette tips is five years
Unit SizeCS