Finntip™ Pipette Tips
Finntip™ Pipette Tips
Actual product may vary
Finntip™ Pipette Tips
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFit for PurposeColor
94020735 mLSterileSterile & PCR ReadyGreen
940030010 μLNon-sterileCertified QualityPink
940031010 μLNon-sterileCertified QualityPink
94010301000 μLNon-sterileCertified QualityBlue
94011101000 μLNon-sterileCertified QualityBlue
94011131000 μLSterileSterile & PCR ReadyBlue
9401250300 μLNon-sterileCertified QualityOrange
9401260300 μLNon-sterileCertified QualityOrange
94020305 mLNon-sterileCertified QualityGreen
94020705 mLNon-sterileCertified QualityGreen
940217110 mLNon-sterileCertified QualityRed
940218010 mLNon-sterileCertified QualityRed
940218310 mLSterileSterile & PCR ReadyRed
940036050 μLNon-sterileCertified QualityTurquoise
940037050 μLNon-sterileCertified QualityTurquoise
Catalog number 9402073
Price (MYR)
723.00
PK
Add to cart
Volume (Metric):
5 mL
Sterility:
Sterile
Fit for Purpose:
Sterile & PCR Ready
Color:
Green
Price (MYR)
723.00
PK
Add to cart
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 250, 300 and 1000 μL
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 5mL, Non-Filtered, Sterile, Rack
Volume (Metric)5 mL
FilteredNon-filtered
SterilitySterile
ColorGreen
ModelFinntip 5mL
Length (English)5.8 in.
Length (Metric)14.7cm
FormatRack
Color CodeGreen
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip
Fit for PurposeSterile & PCR Ready
Unit SizePK