Search Thermo Fisher Scientific
Ensure accurate, precise pipetting. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
| Catalog Number | Sterility | Fit for Purpose | Format |
|---|---|---|---|
| 9400620 | Non-sterile | Certified Quality | Bulk |
| 9400610 | Non-sterile | Certified Quality | Rack |
Accurate, precise pipetting with 384-format volume 20 μL. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
Proven quality
Immediate traceability
Note:
Availability may vary by country.