Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Actual product may vary
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers
Provide uncompromised sample protection for -50° to -86°C applications with Thermo Scientific Forma 900 Series -86°C Upright Ultra-Low Temperature Freezers. These freezers are designed for daily sample protection and dependability.
Have Questions?
Catalog NumberHoldsdimCapacityEnglishDimensions (DxWxH) Exterior
906GP-ULTSBoxes (400)38.9 x 43.4 x 77.9 in.
Catalog number 906GP-ULTS
Price (MYR)
-
Holds:
Boxes (400)
Dimensions (DxWxH) Exterior:
38.9 x 43.4 x 77.9 in.
Provide uncompromised sample protection for -50° to -86°C applications with Thermo Scientific Forma 900 Series -86°C Upright Ultra-Low Temperature Freezers. These freezers are designed for daily sample protection and dependability.
Key Features
  • Centralized, eye level information center, including microprocessor control and monitoring system; all controls and displays are easy to reach and read
  • Improves temperature control and increases reserve BTUH capacity, resulting in a more stable temperature sample
  • Single-hand operation: easy-to-use, padlock-compatible, ergonomic door handle with integrated key lock
  • Optional CO2 or LN2 back-up: Choice of CO2 or LN2 safety back-up systems for additional sample protection in the event of a power or mechanical failure
  • Optional chart recorder: Built-in chart recorder can be installed at eye level on Forma 900 -86°C models
Construction:
  • 5 in. (127mm) foamed-in-place, polyurethane insulation: Reduces power consumption, maintains temperature setpoint
  • Triple-point door gasket, provides longer holding time in the event of a door opening
  • Rugged, heavy gauge, cold-rolled steel cabinets resist chipping and rust
  • 1 in. (25mm) access ports: Allow for the use of inexpedient probes or instrumentation
  • Four inner doors reduce cold air loss and improve temperature recovery after door closings
  • Painted Galvaneel interior (stainless steel option available)
Warranty varies by country, please contact us for details
Specifications
Door StyleInner (4)
No. of Doors4 (Inner)
Cabinet MaterialCold-rolled Steel
InsulationPolyurethane Foam
InteriorStainless Steel
StyleUpright
Dimensions (DxWxH) Exterior38.9 x 43.4 x 77.9 in.
HoldsBoxes (400)
Unit SizeEA
Frequently asked questions (FAQs)
The low battery alarm is displaying on my ULT (Ultra-Low Temperature) freezer. Can I buy a new battery?
You can order a new battery (Cat. No. 400159) online.