Serological Pipets
Serological Pipets
Actual product may vary
Thermo Scientific
Serological Pipets
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
Have Questions?
Catalog NumberVolume (Metric)GraduationsColor
9.40022 mL1/100Green
Catalog number 9.4002
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
2 mL
Graduations:
1/100
Color:
Green
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
  • Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
  • Smooth interiors, prevent cell hang-up or clumping inside the pipette
  • Excellent optical transparency
  • Color coded cotton plugs and dispenser boxes for easy identification
  • Convenient, stackable TPP dispenser box
  • Single wrapped in lint-free paper/plastic blister
  • Slender and short form enables ergonomic working conditions in the sterile hood
  • Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
  • Highly drop free pipet tip design
  • Optimized form of the mouthpiece preserves the rubber of the pipet aid
  • Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Specifications
MaterialPolystyrene
GraduatedYes
Graduations1/100
ColorGreen
Volume (Metric)2 mL
Unit SizeEach