Check Order Status
HiPur™ 10% Neutral Buffered Formalin
Actual product may vary
HiPur™ 10% Neutral Buffered Formalin
Choose the option that best fits your workflow, HiPur™ 10% neutral buffered formalin is available in one and five gallon sizes.
Have Questions?
Catalog Number
Quantity
9.4001
4 x 1 gal.
Catalog number
9.4001
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 gal.
Specifications
Color
Colorless
Concentration
0.1
Quantity
4 x 1 gal.
For Use With (Application)
Histological fixative to fix tissues
Unit Size
Each
Contents & Storage
Room temperature