Check Order Status
Hydrion™ pH Test Paper
Actual product may vary
Hydrion™ pH Test Paper
Micro Essential Laboratory Hydrion™ pH Test Paper is suitable for easy and instant pH determination.
Have Questions?
Catalog Number
pH Range
9.4
1 to 14
Catalog number
9.4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
pH Range:
1 to 14
Wide pH range
Distinct color match at each full pH unit
Immediate response with accurate, reproducible readings
Each kit consists of a 4.57 m (15 ft.) long x 6.35 mm (0.25 in.) wide roll of test paper (100 to 150 tests)
For classrooms, hospital and general laboratories, idustrial testing, food service, safety/emergency testing, agriculture, in vitro testing
Calibration verified using NIST traceable standards
Specifications
pH Range
1 to 14
For Use With (Application)
pH Testing
Quantity
10/Cs.
Type
pH Test Paper
Length (Metric)
4.57 m
Width (Metric)
6.35 mm