MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
Have Questions?
Catalog Number
Quantity
Packaging Type
9.2003
30/Pk.
Individually Packed
Catalog number 9.2003
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
30/Pk.
Packaging Type:
Individually Packed
Ready to use. No preparation or dilution of antimicrobial agent is required. Simply place on an inoculated agar plate and incubate
MIC testing for a wide range of agents and against many organisms can be done on request and at short notice
Synergy Testing: can be crossed for evaluating a critical assay such as the in vitro combination of antibiotics
Rapid and Efficient: easy to set up. Full range of MIC testing of 6 agents on a large plate takes no more than 5 minutes
Solid Medium: Growth of organisms on agar better mimics in vitro conditions. It is easy to visualize hetero-resistant variants in a population and to detect mixed cultures and contamination
Inoculum Tolerant, Macro.Method: a stable gradient technique that is inoculum tolerant (changes in inoculum do not affect the MIC results for a susceptible strain), thus it can be used in a macro-format with heavier inoculum and a richer media to maximize the detection of low-level and hetero-resistance
Easy to Handle: The package is small and light weigh. Required storage space is minimal
Wide MIC Range: spans across 15 dilutions fulfilling pharmacokinetic needs, varying susceptibility of many organism groups, and covers different MIC breakpoints used in different countries as recommended by various reference or expert groups
Easy to Learn: Minimal training is required for clinical microbiology personnel
No equipment is required for use
Easy for New Agents
MIC Equivalent to Reference Values
Improved MIC Precision
Flexible Media Choice: can be used as a quality control tool for checking different AST media, brand to brand and batch to batch
Robust Technique
Generates quantitative MIC values that are needed to guide therapy selection for the individual patient - choice, dose and regimen
Can be used for Fastidious and Slow Growers
Can be used to directly test positive blood cultures, CSF and sputum from cystic fibrosis patients and bronchial aspirates from VAP patients. The extra day saved by testing the specimen directly can have significant impact in therapy management, patient care and hospital cost savings
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.
Specifications
Antimicrobial AgentsAmpicillin
Concentration0.016-256 μg/mL
DescriptionLiofilchem™ MTS™ Ampicillin
Antibiotic CodeAMP
Product TypeMIC Test Strip
For Use With (Application)Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms