910 Stainless Steel/HPPE Cut-Resistant Gloves
910 Stainless Steel/HPPE Cut-Resistant Gloves
Actual product may vary
Thermo Scientific
910 Stainless Steel/HPPE Cut-Resistant Gloves
Flexible and lightweight gloves are available in uncoated or PVC coated styles and are ideal for light-duty applications.
Have Questions?
Catalog NumberSizeCoating Material
9.10066Uncoated
Catalog number 9.1006
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
6
Coating Material:
Uncoated
  • High-performance polyethylene engineered fiber for added strength, comfort and cut resistance
  • Fully launderable and color coded for easy identification
  • Gauge: 10
  • Uncoated or PVC dotted style available
Specifications
Certifications/Compliance (Europe)CE
ConstructionSeamless Knit
Coating MaterialUncoated
ColorWhite
Cut Resistance RatingANSI A4
HandAmbidextrous
MaterialPolyethylene
Product TypeCut-resistant Gloves
Size6
EN388 StandardEN 388:2X43DP
For Use With (Application)Food processing, machine or compartment assembly, bottling, sheet Metal handling, construction, recycling, Glass handling, razor blades
Glove Cuff StyleKnit
Yarn Gauge10
Unit SizeEach