PYREX™ PTFE Plug Only for Craig Tube
Product Image
PYREX™ PTFE Plug Only for Craig Tube
For PYREX 6949M series microchemistry kits
Have Questions?
Catalog number 9.1003
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Specifications
Product TypePlug
For Use With (Equipment)For PYREX™ 6949M series microchemistry kit, replacement part for Craig tube
MaterialPTFE
Length (English)3.34 in.
Length (Metric)85 mm
Diameter (English) Outer0.39 in.
Diameter (Metric) Outer10 mm