Check Order Status
UNISOL pH Indicator Solution
UNISOL pH Indicator Solution
For pH determination in pure water, in surface waters, and in very dilute acids or bases.
Have Questions?
Catalog Number
pH Range
9.1002
4 to 10
Catalog number
9.1002
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
pH Range:
4 to 10
pH determinations in weakly buffered solutions
UNISOL indicator solutions are used for pH determination in pure water, in surface waters and in very dilute acids or bases.
An indicator solution is added to the sample and the reaction color is compared with a color scale.
Therefore, they allow the easy and reliable pH determination in weakly buffered solutions.
Specifications
pH Range
4 to 10
Volume (Metric)
100 mL
Includes
Color chart, cuvette
Quantity
1 Bottle
Shelf Life
3 Years
Type
pH Indicator Solution