CO₂ Resistant Shakers and Packages
CO₂ Resistant Shakers and Packages
Actual product may vary
CO₂ Resistant Shakers and Packages
CO₂ Resistant Shakers and Packages
CO₂ Resistant Shakers and Packages
Unlike a combined shaking incubator, a CO2 resistant shaker can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your samples
Have Questions?
Catalog NumberIncludes
88881104BCO2 resistant shaker 3 mm orbit, CN, UK, EU style plugs 220-240 V including control box with 4-20 mA data output, 14.1 x 11.8 in universal platform (360 X 300 mm), and flask clamp starter kit
Catalog number 88881104B
Price (MYR)
24,703.00
EA
Add to cart
Includes:
CO2 resistant shaker 3 mm orbit, CN, UK, EU style plugs 220-240 V including control box with 4-20 mA data output, 14.1 x 11.8 in universal platform (360 X 300 mm), and flask clamp starter kit
Price (MYR)
24,703.00
EA
Add to cart
Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO2 Resistant Shakers.
  • Orbital shaking through magnetic power eliminates the need to replace belts
  • Small footprint ideal for use in a CO2 incubator
  • Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions
  • Convenient external control box with LED display allows for easy adjustments without opening the chamber door
  • Minimal heat dissipation assures conditions within the CO2 chamber are not effected
  • Stainless steel construction for protection from corrosive CO2 gassing conditions
  • An extensive selection of accessories fits most applications
  • CO2 resistant shaker can be used outside of CO2 incubator for maximum value
  • External control box for ease-of-use without disrupting your samples
  • Comes standard with 4-20 mA data output port to provide added security and remote monitoring capability
A large selection of available accessories to fit most applications:
  • Wide variety of flask clamps available
  • Fixed and adjustable tube racks available
  • 96-well microplate holder
  • Rubber and sticky mats
  • Flask clamp starter kit for ease and flexibility
  • Large capacity 18x18 in. universal platform
Applications:
  • Mammalian cell culture
  • Bacterial suspensions
  • Staining and destaining
  • Washing procedures
  • Hybridization
  • Solubility studies
  • Extraction procedures
Warranty:
  • 2 year Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications
Orbit3 mm
Load Bearing Capacity (Metric)6 kg
Speed Range30 to 1200 rpm, ±1 rpm
Voltage100/240 V
Temperature Range (Metric)4°C to 60°C
Certifications/CompliancecCSAus, CE Mark, RoHS, WEEE, FCC, CA 65, REACH, ICES-001
Temperature Range (English)39.2°F to 140°F
Controller TypeCO2 Resistant
ModelCO2 Resistant Shaker
IncludesCO2 resistant shaker 3 mm orbit, CN, UK, EU style plugs 220-240 V including control box with 4-20 mA data output, 14.1 x 11.8 in universal platform (360 X 300 mm), and flask clamp starter kit
Unit SizeEA