CO2 Resistant Shakers
CO<sub>2</sub> Resistant Shakers
Actual product may vary
CO2 Resistant Shakers
Benefit from reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment.
Have Questions?
Catalog NumberIncludesPlug Type
88881102Shaker, control box, plugsEU, UK, CN Plug
Catalog number 88881102
Price (MYR)
25,232.00
EA
Add to cart
Includes:
Shaker, control box, plugs
Plug Type:
EU, UK, CN Plug
Price (MYR)
25,232.00
EA
Add to cart
Get reliable around-the-clock operation ideally suited to keep your cells alive and flourishing within your working environment with Thermo Scientific™ CO2 Resistant Shakers.
  • Orbital shaking through magnetic power eliminates the need to replace belts
  • Small footprint ideal for use in a CO2 incubator
  • Specially treated mechanical components protect up to 20% CO2 and 95% humidity conditions
  • Convenient external control box with LED display allows for easy adjustments without opening the chamber door
  • Minimal heat dissipation assures conditions within the CO2 chamber are not effected
  • Stainless steel construction for protection from corrosive CO2 gassing conditions
  • An extensive selection of accessories fits most applications
  • CO2 resistant shaker can be used outside of CO2 incubator for maximum value
  • External control box for ease-of-use without disrupting your samples
A large selection of available accessories to fit most applications:
  • Wide variety of flask clamps available
  • Fixed and adjustable tube racks available
  • 96-well microplate holder
  • Rubber and sticky mats
  • Flask clamp starter kit for ease and flexibility
Applications:
  • Mammalian cell culture
  • Bacterial suspensions
  • Staining and destaining
  • Washing procedures
  • Hybridization
  • Solubility studies
  • Extraction procedures
Warranty:
  • 2 year Thermo Fisher Scientific standard limited warranty. See your sales representative for details.
Specifications
Orbit19 mm
Load Bearing Capacity (Metric)6 kg
Speed Control Accuracy1 rpm
Speed Range30 to 300 rpm, ±1 rpm
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Temperature Range (Metric)4°C to 60°C
Product TypeCO2 Resistant Shaker
Load Bearing Capacity (English)13 lb.
Certifications/CompliancecCSAus, CE Mark, RoHS, WEEE
Plug TypeEU, UK, CN Plug
Temperature Range (English)39.2°F to 140°F
Frequency50/60 Hz
Dimensions (L x W x H)11.8 x 14.2 x 3.8 in. (300 x 360 x 96 mm)
Weight (English)29 lb.
Weight (Metric)13 kg
Controller TypeCo2-Resistant
DescriptionCO2 Resistant Shaker
ModelCO2 Resistant Shaker
DisplayLED, speed and time
IncludesShaker, control box, plugs
Timer Range0 to 99:59 hr., Continuous
Unit SizeEA