Accessories for Digital Heating Cooling Drybath
Accessories for Digital Heating Cooling Drybath
Actual product may vary
Accessories for Digital Heating Cooling Drybath
Optimize the performance of our digital heating cooling drybath and digital heating shaking bath by using these accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of PlacesTube SizeType
88880133650mL largeBlock
88880130301.5mLBlock
888801311515mLBlock
Catalog number 88880133
View Price:Sign in to see your account pricing. Need an account? Register with us today.
No. of Places:
6
Tube Size:
50mL large
Type:
Block
Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.
  •  Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)50 mL
LightingLED
No. of Places6
TypeBlock
Tube Size50mL large
Dimensions (L x W x H)3 x 3.9 x 3.4 in. (76.5 x 98 x 87 mm)
Unit SizeEA