RT Touch Series Magnetic Stirrers
RT Touch Series Magnetic Stirrers
Actual product may vary
RT Touch Series Magnetic Stirrers
Stay in control with our low-profile, digital display stirrers, ideal for routine stirring applications where visible stir speed is required.
Have Questions?
Catalog NumberCapacitySpeedPlug Type
888800144 L30 to 2000 rpmCord set w/various plugs
Catalog number 88880014
Price (MYR)
2,684.00
EA
Add to cart
Capacity:
4 L
Speed:
30 to 2000 rpm
Plug Type:
Cord set w/various plugs
Price (MYR)
2,684.00
EA
Add to cart
Stay in control with Thermo Scientific™ RT Touch Series Magnetic Stirrers. These low-profile, digital display stirrers are ideal for routine applications where visible stir speed is required.
Strong magnetic coupling ensures consistent stirring during experimental procedures. Gradual start-up stirring prevents splashing.
  • Digital control
  • Designed for stir rod support
  • Low-profile, lightweight design with small footprint
  • Available in two sizes
  • Speed control: 30 to 2000rpm
Includes:
Two nonslip silicone plate covers
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed30 to 2000 rpm
Top Plate MaterialPolypropylene
Capacity4 L
DisplayDigital
Voltage100/240 V
Wattage6 w
Dimensions (L x W x H)11.02 x 8.27 x 2.82 in. (280 x 210 x 72 mm)
Height (English)2.82 in.
Height (Metric)72 mm
Length (English)11.02 in.
Length (Metric)280 mm
Top Plate Diameter (English)6.69 in.
Top Plate Diameter (Metric)170 mm
Width (English)8.27 in.
Width (Metric)210 mm
Electrical Requirements100 to 240V 50/60Hz
Frequency50/60 Hz
Plug TypeCord set w/various plugs
Certifications/ComplianceCE
Warranty and Service Offering2 Years
TypeStirrer
Unit SizeEA