Block, 6 x 1.5 mL
Product Image
Block, 6 x 1.5 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Have Questions?
Catalog NumberNo. of Places
888711036
Catalog number 88871103
Price (MYR)
189.00
EA
Add to cart
No. of Places:
6
Price (MYR)
189.00
EA
Add to cart
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Description6 x 1.5mL block
Capacity (Metric)1.5 mL
For Use With (Equipment)Thermo Scientific™ Compact Dry Bath/Block Heater
LightingLED
No. of Places6
TypeDry Block
Dimensions (L x W x H)36 x 54 x 30 mm (1.4 x 2.1 x 1.2 in.)
Unit SizeEA