Compact Digital Dry Bath/Block Heater
Compact Digital Dry Bath/Block Heater
Actual product may vary
Compact Digital Dry Bath/Block Heater
Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelPlug Type
88871004Compact Dry Bath DCN, EU, UK
88871003Compact Dry Bath SCN, EU, UK
Catalog number 88871004
Price (MYR)
3,799.00
EA
Add to cart
Model:
Compact Dry Bath D
Plug Type:
CN, EU, UK
Price (MYR)
3,799.00
EA
Add to cart
Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
Required Accessory:
  • Unit must be ordered with block(s)
Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, cTUVus, RoHS
Controller TypeDigital
Capacity (Metric)0.2 mL
LightingLED
Plug TypeCN, EU, UK
Temperature Range (Metric)Ambient +2° to 100°C
TypeDry Block Heater
Heating Rate20 min. average heating rate (30° to 100°C, ambient at 25°C)
IncludesDry bath, CN, EU,and UK style plugs, block lifter, plastic lid
ModelCompact Dry Bath D
No. of Blocks2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 0.5°C
Warranty1 Year, Replacement
Dimensions (L x W x H)200 x 200 x 25 mm (7.9 x 7.9 x 1 in.)
Length (English)7.9 in.
Length (Metric)200 mm
Width (English)7.9 in.
Width (Metric)200 mm
Height (English)1 in.
Height (Metric)25 mm
Weight (English)3.3 lb.
Weight (Metric)<1.5 kg
Electrical Requirements12 V, DC, 7 A
Voltage100/240 V
Unit SizeEA