Block, 0.5 mL and 0.2 mL
Block, 0.5 mL and 0.2 mL
Actual product may vary
Block, 0.5 mL and 0.2 mL
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberNo. of Places
8887011530
Catalog number 88870115
HSN 39269099
Price (MYR)
1,332.00
EA
Add to cart
No. of Places:
30
Price (MYR)
1,332.00
EA
Add to cart
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
DescriptionMixed block, 0.5mL and 0.2mL
Capacity (Metric)30 x 0.5 mL, 20 x 0.2 mL
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds30 x 0.5mL and 20 x 0.2mL
LightingLED
No. of Places30
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit SizeEA