Search Thermo Fisher Scientific
| Catalog Number | For Use With (Equipment) |
|---|---|
| 850-WT | Waters Columns Replacement Tips |
| 850-00 | 4.6 mm I.D. Guard Columns |
| 852-00 | 2.1 to 3 mm I.D. Guard Columns |
Eliminate the requirement for extra fittings using Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders. They are reusable, stainless-steel guard cartridge holders that attach directly to the analytical column inlet.
Ordering Information:
Other column dimensions are available. Please call your Customer Service Representative for more information.