Check Order Status
AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration
Actual product may vary
AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Have Questions?
Catalog Number
Pore Size
Volume (Metric)
Material (Membrane)
8127
also known as 8127CE
30 to 40 μm
1 mL
Polypropylene/Polyethylene Non-woven Media
Catalog number
8127
also known as 8127CE
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
30 to 40 μm
Volume (Metric):
1 mL
Material (Membrane):
Polypropylene/Polyethylene Non-woven Media
Plates are constructed of chemically resistant, biologically-inert polypropylene
This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Material (Membrane)
Polypropylene/Polyethylene Non-woven Media
Pore Size
30 to 40 μm
Sterility
Non-sterile
Housing Material
Polypropylene Housing
Quantity
5/Pk.
Volume (Metric)
1 mL