LN2 Supply Tanks
LN<sub>2</sub> Supply Tanks
Actual product may vary
LN2 Supply Tanks
Transport cryogenic industrial gases with these LN2 Supply tanks, which are designed to be both durable and user-friendly.
Have Questions?
Catalog NumberCapacity (Metric)
8121230 L
Catalog number 8121
Price (MYR)
75,468.00
EA
Add to cart
Capacity (Metric):
230 L
Price (MYR)
75,468.00
EA
Add to cart
Transport cryogenic industrial gases with Thermo Scientific™ LN2 Supply tanks, which are designed to be both durable and user-friendly.
For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilizing controlled-rate freezing equipment.
  • Rated at 22psi (151.7kPa)
  • Evaporate 2% of volume per day
  • Warranty varies by country, please contact us for details
Specifications
TypeLN2 Supply Tank
Capacity (English)8.122 cu. ft.
Capacity (Metric)230 L
Height (English) Exterior54.75 in.
Height (Metric) Exterior139.2 cm
Diameter (English)26 in.
Diameter (Metric)66 cm
Unit SizeEA