Aquasil™ 5μm C18 HPLC Columns, 5?m Particle Size
Aquasil™ 5μm C18 HPLC Columns, 5?m Particle Size
Actual product may vary
Aquasil™ 5μm C18 HPLC Columns, 5?m Particle Size
Thermo ScientificTM AquasilTM 5μm C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
77505-254630
also known as 77505-254630B
250 mm4.6 mm
77505-254030
also known as 77505-254030B
250 mm4 mm
Catalog number 77505-254630
also known as 77505-254630B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Thermo ScientificTM AquasilTM 5μm C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
  • Polar endcapped C18 phase
  • Alternative selectivity compared to conventional C18 phases
  • Compatible with 100% aqueous mobile phases
  • Ordering Information:
    Other column dimensions are also available. Please call Customer Service for more information.
    Specifications
    Column FormatAnalytical Column
    Diameter (Metric)4.6 mm
    For Use WithLC/MS
    Length (Metric)250 mm
    Particle Size5 μm
    pH2 to 8
    Pore Size100 Å
    Surface Area310 m²/g
    USP TypeL1
    Stationary PhaseC18
    Packing MaterialSilica
    Carbon Load12%
    Column TypeReversed Phase
    EndcappedYes
    PhaseReversed Phase
    Unit SizeEach