Fiberlite™ F14-6 x 250LE Rotor Adapters
Fiberlite™ F14-6 x 250LE Rotor Adapters
Actual product may vary
Fiberlite™ F14-6 x 250LE Rotor Adapters
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
75100136
also known as LT75100136
50 mL Conical Tube (Set of 2)
010-013850 mL Round-bottom Tube (Set of 2)
Catalog number 75100136
also known as LT75100136
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
50 mL Conical Tube (Set of 2)
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:
  • Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.
Specifications
TypeRotor Adapter
Tube TypeConical
Capacity50 mL Conical Tube (Set of 2)
Tube Size50 mL
Unit SizeEA