Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Actual product may vary
Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Medifuge™ Small Benchtop Centrifuge
Perform both routine clinical and life science separations with this compact centrifuge and the unique DualSpin rotor featuring a 2-in-1 hybrid design.
Have Questions?
Catalog NumberElectrical RequirementsDescription
75008800220/230 V 50/60 HzMedifuge Centrifuge
Catalog number 75008800
Price (MYR)
-
Electrical Requirements:
220/230 V 50/60 Hz
Description:
Medifuge Centrifuge
Perform both routine clinical and life science separations with the Thermo Scientific™ Medifuge™ Clinical Centrifuge. It fits in with its compact design and stands out with the unique Thermo Scientific™ DualSpin™ rotor featuring a 2-in-1 hybrid design. Easily change between fixed angle and swinging buckets using the same rotor. Four customizable programs allow you to quickly run routine protocols. Three selectable deceleration profiles, including brake-off option, are designed to optimize separation.
Package content
  • Each Medifuge centrifuge comes as a package, including:
  • Medifuge small benchtop centrifuge
  • DualSpin rotor body
  • Fixed angle buckets (set of 8)
  • Swinging buckets (set of 8)
  • Green/short spacers (set of 8)
  • Yellow/long spacers (set of 8)
Applications
  • Platelet-poor plasma (PPP) separation
  • Urinalysis
  • Clinical chemistry analysis
  • Hematology analysis
  • Serum preparation using gel tube
  • Cell culture pelleting
Features
  • Unique DualSpin rotor with interchangeable fixed angle and swinging buckets accommodates 1.4 mL to 15 mL tubes with two spacer options
  • The lightweight composite 8-place rotor is easy to handle and saves energy during acceleration/deceleration
  • Large, brightly-lit display is easy to read and fast one-click lid closure simplifies operation
  • Four customizable programs to quickly run routine protocols
  • Three selectable deceleration profiles including brake-off option are designed to optimize separation
  • Quiet operation at noise level less than 56 dBA
  • Emergency lid lock release available in case of power failure
Certifications
UL, CE, RoHS, CSA, IVD, FDA class 1
Standards
IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, EN ISO 13485
Previous models
  • Labofuge centrifuge
  • Medilite centrifuge
Specifications
Capacity8 x 15 mL (Fixed Angle), 8 x 10 mL (Swinging Bucket)
Max. Speed4,900 rpm
Max. RCF3,114 x g (Fixed Angle); 3,490 x g (Swinging Bucket)
RefrigeratedNo
DisplayLarge, Bright LED
Height (English) Exterior94.3 in.
Height (Metric) Exterior240 mm
Length (English) Exterior176.8 in.
Length (Metric) Exterior450 mm
Noise Level<56 dB(A) (swinging bucket)
Safety FeaturesLid lock, steel armored ring, view port for speed calibration, lid emergency release
Width (English) Exterior127.7 in.
Width (Metric) Exterior325 mm
Wattage130 w
Certifications/ComplianceUL, CE, RoHS, CSA, IVD, FDA class 1
IncludesDualSpin rotor body, fixed angle buckets (set of 8), swinging buckets (set of 8), green/short spacers (set of 8), yellow/long spacers (set of 8)
DescriptionMedifuge Centrifuge
Electrical Requirements220/230 V 50/60 Hz
Controller TypeMicroprocessor
Dimensions (HWD)9.5 x 12.8 x 17.7 in. (24 x 32.5 x 45 cm)
Drive SystemInduction drive
Net Weight34.1 lb. (15.5 kg)
Program StorageYes, 4 program memory
Run Time1-99 minutes, in increments of 1 minute; HOLD
Temperature Range2°C to 40°C
Product LineMedifuge
Height (English) Exterior Lid Open200.4 in.
Profile (Acceleration/Braking)1 accel (standard)/3 decel (standard, soft, brake-off)
Frequency50/60 Hz
Height (Metric) Exterior Lid Open510 mm
Voltage220/230 V
Unit SizeEA