Check Order Status
Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
Actual product may vary
Thermo Scientific
Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
Have Questions?
Change view
Catalog Number
Color
75008188
Gray
75008183
Green/Yellow
75008182
Green
75008187
Brown
4 Options
Catalog number
75008188
Price (MYR)
-
Color:
Gray
Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Specifications
For Use With (Equipment)
4 x 180 Swing bucket rotor
Color
Gray