Check Order Status
Round Bucket for Thermo Scientific™ Sorvall™ Stratos Rotor
Actual product may vary
Round Bucket for Thermo Scientific™ Sorvall™ Stratos Rotor
For use with Four-Place Swinging Bucket Rotor 75-003-047
Have Questions?
Catalog number
75008172
HSN
84219100
Price (MYR)
0.00
EA
Add to cart
Price (MYR)
0.00
EA
Add to cart
Specifications
For Use With (Equipment)
Swing out rotor for Labofuge 400 and 400R centrifuge
Capacity
18 mL
Tube Size
1.5 to 100 mL