4 x 100mL Swinging Bucket Rotor
4 x 100mL Swinging Bucket Rotor
Actual product may vary
4 x 100mL Swinging Bucket Rotor
Combine Thermo Scientific™ 4 x 100mL Swinging Bucket Rotor with an optional aerosol-tight cap for added sample protection.
Have Questions?
Catalog NumberCapacity
750075914 x 100 mL
Catalog number 75007591
Price (MYR)
-
Capacity:
4 x 100 mL
Combine the Thermo Scientific™ 4 x 100mL Swinging Bucket Rotor with an optional aerosol-tight cap for extra protection.
Specifications
  • Capacity is flexible with adapters: 1.5 to 100mL
  • Capacity: 4 × 100mL
  • Maximum speed/RCF: 4000rpm/2590 x g
Ordering Information:
Order round buckets (75007555), tube adapters and aerosol-tight cap separately. Contact your service representative for information.
Compatible with:
Thermo Scientific™ Primo™/Primo R
Specifications
Description4 x 100mL Swinging Bucket Rotor
TypeSwinging Bucket Rotor
Capacity4 x 100 mL
Max. RCF2,590 x g
Max. Speed4,000 rpm
Radius (Metric)14.5 cm
Max. Tube Length100 mm
Max. Tube Diameter44 mm
For Use With (Equipment)Thermo Scientific Sorvall Primo and Primo Centrifuges
Certifications/ComplianceBiocontainment certified by CAMR, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA