Check Order Status
4-Place Swinging Bucket Rotor Adapters
Actual product may vary
4-Place Swinging Bucket Rotor Adapters
For added processing versatility
Have Questions?
Change view
Catalog Number
Tube Type
75007556
50 mL Falcon Tube
75007544
also known as 75007544FS
15 mL Blood Tube
2 Options
Catalog number
75007556
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Tube Type:
50 mL Falcon Tube
For added processing versatility
Maximum tube size: 30 x 117mm
Specifications
Tube Type
50 mL Falcon Tube
For Use With (Equipment)
Swinging bucket rotor