Adapters for TX-1000 Swinging Bucket Rotor
Adapters for TX-1000 Swinging Bucket Rotor
Actual product may vary
Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
7500730696 x 15 mL15mL Conical Tube
75003829150 to 300 mL Small Blood BagsSmall bag/cell culture bags 4 x 2 bags (< 350mL)
750073014 x 1000 mL1000mL Bio-Bottle
7500425248 x 50 mL50mL Thermo Scientific Nalgene Oak Ridge Tube
750042534 x 500 mL500mL Nalgene Bottle
750053924 x 250 mL250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
750073024 x 500 mL500mL Corning Bottle
750073034 x 6 mL Standard or 4 x 2 mL Deep-WellMicroplate carriers
750073044 x 750 mL750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
750073058 x 250 mL250mL Thermo Scientific Nalgene Bottle
Catalog number 75007306
Price (MYR)
-
Capacity:
96 x 15 mL
For Use With (Equipment):
15mL Conical Tube
An assortment of adapters for outstanding flexibility
Specifications
TypeRotor Adapter
Tube TypeConical
Capacity96 x 15 mL
Max. Tube Diameter17.5 mm
Max. Tube Length121 mm
For Use With (Equipment)15mL Conical Tube
Tube Size15 mL
Unit SizeEA