LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Actual product may vary
LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Support a diverse array of sample processing applications without the need to modify existing protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75006476
also known as R75006476
Swinging Bucket Rotor
75006421Sealing Cap
75006436Double Blood Bag Bucket
Catalog number 75006476
also known as R75006476
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Swinging Bucket Rotor
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Cryofuge 5500i series centrifuges
Specifications
DescriptionLH-4000W Windshield Swinging Bucket Rotor
MaterialStainless Steel Rotor Body, Aluminum Bucket
TypeSwinging Bucket Rotor
Capacity8 x 500 mL
Max. RCF5,480 x g
Max. Speed4,400 rpm
Radius (Metric)2215 mm
Radius (Metric) Min.88 mm
For Use With (Equipment)Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Angle90°
Min. RCF1,903 x g
BiocontainmentYes
Unit SizeEA