Hematocrit Fixed-Angle Rotor
Hematocrit Fixed-Angle Rotor
Actual product may vary
Hematocrit Fixed-Angle Rotor
Hematocrit Fixed-Angle Rotor
Hematocrit Fixed-Angle Rotor
Save time in clinical processing applications with a fixed-angle rotor that provides dedicated blood processing to achieve 24 capillary separations per run.
Have Questions?
Catalog NumberCapacity
7500573324 Capillary Tubes
Catalog number 75005733
HSN 84219100
Price (MYR)
0.00
EA
Add to cart
Capacity:
24 Capillary Tubes
Price (MYR)
0.00
EA
Add to cart
Save time in clinical processing applications with the Thermo Scientific™ Hematocrit Fixed-Angle Rotor, which provides dedicated blood processing to achieve 24 capillary separations per run.
Achieve red blood cell capillary separations.
Result reading chart provided.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Includes linear scale. Replacement capillaries (75000923), sealing kit (75000964) and replacement linear scale (75000938) are sold separately. Contact your sales representative for more information.
Specifications
DescriptionHematocrit Fixed-Angle Rotor
TypeFixed Angle Rotor
Capacity24 Capillary Tubes
Max. RCF17,860 x g
Max. Speed13,300 rpm
Radius (Metric)85 mm
K Factor1559
For Use With (Equipment)Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle90°
Unit SizeEA