Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
Actual product may vary
Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
75005730ClickSeal Biocontainment LidMicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
75003405O-ringMicroClick 24 x 2 Rotor Lid
75005726O-ringMicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Catalog number 75005730
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
ClickSeal Biocontainment Lid
For Use With (Equipment):
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
Specifications
TypeClickSeal Biocontainment Lid
For Use With (Equipment)MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Unit SizeEA