MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Actual product may vary
MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
MicroClick 30 x 2 Fixed Angle Microtube Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Perform micro-volume applications such as nucleic acid preparation at up to 14,000rpm/21,693 x g with the Thermo Scientific™ MicroClick 30 x 2 Fixed Angle Microtube Rotor.
Have Questions?
Catalog NumberCapacity
7500571930 x 2 mL
Catalog number 75005719
Price (MYR)
-
Capacity:
30 x 2 mL
Perform micro-volume applications such as nucleic acid preparation at up to 14,000rpm/21,694 x g with the Thermo Scientific™ MicroClick 30 x 2 Fixed Angle Microtube Rotor.
Maximize capacity for 2mL microtubes with this rotor, designed with an extended neck to accommodate 30 nuclueic acid filter columns.
With Auto-Lock rotor exchange, switch from swing-out to a microtube rotor in as little as 3 seconds with:
  • Trouble-free rotor installation and removal:
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
Coupled with the security of Thermo Scientific™ ClickSeal™ biocontainment lids, certified by Public Health England, Porton Down, UK.
Ordering Information:
Replacement biocontainment lid (75005730) and replacement O-rings (75005726) and adapters are sold separately. Contact your sales representative for more information.
Specifications
DescriptionMicroClick 30 x 2 Fixed Angle Microtube Rotor
TypeFixed Angle Rotor
Capacity30 x 2 mL
Max. RCF21,693 x g
Max. Speed14,000 rpm
Radius (Metric)99 mm
K Factor543
Max. Tube Length45 mm
Max. Tube Diameter11 mm
For Use With (Equipment)Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle45°
BiocontainmentYes
Unit SizeEA