TX-400 4 x 400 mL Swinging Bucket Rotor
TX-400 4 x 400 mL Swinging Bucket Rotor
Actual product may vary
TX-400 4 x 400 mL Swinging Bucket Rotor
Support a wide range of sample sizes and applications with the swinging bucket rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75003655Bucket
75003181Swinging Bucket Rotor
75003656Bucket Lid
Catalog number 75003655
Price (MYR)
-
Type:
Bucket
Support a wide range of sample sizes and applications with the Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor.
Ordering Information:
Buckets and lids are sold separately; order rotor body (75003181) and components to complete assembly. For replacement O-rings for lids (set of four), order 75003657. Tube adapters are also available separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Heraeus™ Multifuge™ X1, Heraeus Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Note:
Rotor 75003181 is the replacement for rotor 75003629
Specifications
TypeBucket
Capacity4 x 400 mL
For Use With (Equipment)TX-400 Rotor
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes
QuantitySet of 4
Unit SizeEA