BIOShield™ 720 High Speed Swinging-Bucket Rotor
BIOShield™ 720 High Speed Swinging-Bucket Rotor
Actual product may vary
BIOShield™ 720 High Speed Swinging-Bucket Rotor
BIOShield™ 720 High Speed Swinging-Bucket Rotor
BIOShield™ 720 High Speed Swinging-Bucket Rotor
BIOShield™ 720 High Speed Swinging-Bucket Rotor
Safely spin frequently-used tubes with the Thermo Scientific™ BIOShield™ 720 High-Speed Swinging-Bucket Rotor.
Have Questions?
Catalog NumberModel
75003183BIOShield 720 Swinging Bucket Rotor w/Slide Coat anti-friction finish
Catalog number 75003183
Price (MYR)
-
Model:
BIOShield 720 Swinging Bucket Rotor w/Slide Coat anti-friction finish
Safely spin frequently-used tubes with the Thermo Scientific™ BIOShield™ 720 High-Speed Swinging-Bucket Rotor.
Ordering Information:
Tube adapters, sold in sets of four, are sold separately. Contact your sales representative for more information.
Compatible with:
Thermo Scientific™ Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Specifications
TypeSwinging Bucket Rotor
Capacity4 x 180 mL
Max. RCF7,188 x g
Max. Speed6,300 rpm
Radius (Metric)162 mm
Max. Tube Length122 mm
Max. Tube Diameter50 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Angle90°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
ModelBIOShield 720 Swinging Bucket Rotor w/Slide Coat anti-friction finish
Unit SizeEA