Check Order Status
Replacement Sealing O-Rings for HIGHConic™ II Fixed-Angle Rotors
Actual product may vary
Replacement Sealing O-Rings for HIGHConic™ II Fixed-Angle Rotors
Replace cracked, dry or otherwise faulty O-rings with this replacement set of two that includes grease.
Have Questions?
Catalog Number
For Use With (Equipment)
75003058
HiGhConic II Fixed-angle Rotor
Catalog number
75003058
Price (MYR)
-
For Use With (Equipment):
HiGhConic II Fixed-angle Rotor
Replace cracked, dry or otherwise faulty O-rings with Thermo Scientific™ Replacement Sealing O-Rings, a replacement set of two that includes grease.
Specifications
Type
O-ring
For Use With (Equipment)
HiGhConic II Fixed-angle Rotor
Description
Replacement Sealing O-rings