A27-8 x 50 Fixed Angle Rotor
A27-8 x 50 Fixed Angle Rotor
Actual product may vary
A27-8 x 50 Fixed Angle Rotor
Shorten run set-up time while providing peace of mind that the rotor is secure with Thermo Scientific™ A27-8 x 50mL Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
750030088 x 50 mL
Catalog number 75003008
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
8 x 50 mL
Shorten run set-up time while providing peace of mind that the rotor is secure with Thermo Scientific™ A27-8 x 50mL Fixed-Angle Rotor. Featuring high RCF performance, this rotor enables virus, proteomics and subcellular particle separations in a modernized Thermo Scientific SS-34 design.
Auto-Lock Rotor Exchange:
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionA27-8 x 50 Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity8 x 50 mL
Max. RCF87,207 x g
Max. Speed27,000 rpm
Radius (Metric)107 mm
Radius (Metric) Min.33 mm
K Factor408
Max. Tube Length108 mm
Max. Tube Diameter29 mm
Net Weight15.0 lb. (6.8 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle34°
BiocontainmentYes
Unit SizeEA