BIOFlex™ HC Swinging-Bucket Rotor
BIOFlex™ HC Swinging-Bucket Rotor
Actual product may vary
BIOFlex™ HC Swinging-Bucket Rotor
Process more than 35 tube styles with the Thermo Scientific™ BIOFlex™ HC Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
750030004 x 1000 mL
Catalog number 75003000
Price (MYR)
67,529.00
EA
Add to cart
Capacity:
4 x 1000 mL
Price (MYR)
67,529.00
EA
Add to cart
Process more than 35 tube styles—from microtubes to 1L bottles, plus conical tubes, microplates, tissue culture flasks and blood bags—with the Thermo Scientific™ BIOFlex™ HC Swinging-Bucket Rotor. Innovations such as Auto-Lock rotor exchange, Auto-ID instant rotor identification and speed handles on each rotor lid shorten run set-up time while providing peace of mind that the rotor is secure.
Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionBIOFlex HC Swinging Bucket Rotor
MaterialStainless Steel Rotor Body, Aluminum Bucket
TypeSwinging Bucket Rotor
Capacity4 x 1000 mL
Max. RCF7,068 x g
Max. Speed5,500 rpm
Radius (Metric)209 mm
Radius (Metric) Min.108 mm
K Factor5,522
Max. Tube Length140 mm
Max. Tube Diameter126 mm
Net Weight11.8 lb. (5.35 kg) Rotor Cross, 22.5 lb. (10.2 kg) Cross with Buckets
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
AngleHorizontal
BiocontainmentYes
Unit SizeEA