BetaBasic™ Phenyl Columns, 5μm Particle Size
BetaBasic™ Phenyl Columns, 5μm Particle Size
Actual product may vary
BetaBasic™ Phenyl Columns, 5μm Particle Size
BetaBasic columns are highly efficient and reproducible for small molecules, peptides and protein digests
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
71805-254630250 mm4.6 mm
HYP71805-254630250 mm4.6 mm
Catalog number 71805-254630
Price (MYR)
-
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Film Thickness5 μm
For Use WithLC/MS Separation
Functionality/Interaction TypeStable bonding
Length (Metric)250 mm
Particle ShapeSpherical
Particle Size5 μm
Pore Size150 Å
Surface Area200 m²/g
USP TypeL11
Stationary PhasePhenyl (Ph)
Product LineBeta
Packing MaterialSilica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeColumn
Unit SizeEach