Restricted Area Signs
Restricted Area Signs
Actual product may vary
Thermo Scientific
Restricted Area Signs
Restricted Area safety signs from Brady™ help to manage and sustain a clean and safe workplace. They are available in a variety of materials and sizes to best fit your application.
Have Questions?
Catalog NumberMaterialDisplayed Text
71389Fiberglass, PolyesterRestricted Area Unauthorized Persons Keep Out
Catalog number 71389
Price (MYR)
-
Material:
Fiberglass, Polyester
Displayed Text:
Restricted Area Unauthorized Persons Keep Out
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Antigenphospho-Smad3 (Ser423/425)
Certifications/ComplianceISO 9001, ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646
ApplicationsWestern Blot
ClassificationPolyclonal
ConjugateUnconjugated
MaterialFiberglass, Polyester
Quantity1 Ea.
FormulationUnpurified rabbit serum in buffer containing 0.05% sodium azide.
Tensile Strength9000 psi
Adhesive TypeNon-adhesive
DescriptionFiberglass Reinforced Polyester
Gene SymbolsDKFZp586N0721; DKFZp686J10186; HSPC193; HsT17436; JV15-2 ; MADH3; MGC60396; Mad3 ; hMAD-3; hSMAD3
Host SpeciesRabbit
ImmunogenKLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Ser423/425 of human Smad3.
Mounting TypeSurface Mount
For Use With (Application)Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs and equipment warning signs, recommended for both indoor and outdoor use
Purification MethodUnpurified
Height (English)10 in.
Quantity100 μL
Max. Temperature (English)193°F
Min. Temperature (English)-40°F
Primary or SecondaryPrimary
Gene ID (Entrez)NM_005902
Thickness0.1 in.
Width (English)14 in.
Width (Metric)35.56 cm
Temperature Range (Metric)-40°C to 90°C
Target SpeciesHuman, Mink, Mouse, Rat
Temperature Range (English)-40°F to 193°F
Dimensions (L x W)10 x 14 in.
Displayed TextRestricted Area Unauthorized Persons Keep Out
Product TypeAdmittance Sign
ColorRed on White
Unit SizeEach
Contents & Storage
-20°C in undiluted aliquotes for up to 24 months, Avoid Freeze/Thaw Cycles