Check Order Status
Silver Nitrate, 0.0500 N (N/20), Ricca Chemical
Element Search
Structure Search
Silver Nitrate, 0.0500 N (N/20), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
69701
4 L
Black Poly Bottle
Catalog number
69701
Price (MYR)
-
Quantity:
4 L
Packaging:
Black Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Silver Nitrate
CAS
7761-88-8
CAS Min %
97.0
Column Format
Guard Cartridge, 2 Pk.
CAS Max %
100.0
View more