Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Actual product may vary
Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Have Questions?
Catalog NumberFor Use With (Equipment)Type
669623.3, 45.8, 51.1, 78.8 cu. ft. UnitsOpen Wire Shelves
Catalog number 6696
HSN 84189900
Price (MYR)
939.00
EA
Add to cart
For Use With (Equipment):
23.3, 45.8, 51.1, 78.8 cu. ft. Units
Type:
Open Wire Shelves
Price (MYR)
939.00
EA
Add to cart
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
  • Warranty varies by country, please contact us for details
Specifications
TypeOpen Wire Shelves
Exterior FinishEpoxy-coated Steel
For Use With (Equipment)23.3, 45.8, 51.1, 78.8 cu. ft. Units
WarrantyWarranty varies by country, please contact us for details
HoldsGeneral Freezer Items
Unit SizeEA