Check Order Status
IC 261
Actual product may vary
IC 261
CK1 inhibitor; displays selectivity for CK1δ and CK1ε
Have Questions?
Catalog Number
Quantity
66765
5 mg
Catalog number
66765
Price (MYR)
-
Quantity:
5 mg
ATP-competitive casein kinase 1 (CK1) inhibitor
Displays >15 fold selectivity for CK1δ and CK1ε over CK1α (IC50 values are approximately 1 μM for CK1δ and CK1ε and 16 μM for CK1α)
Arrests cell cycle at postmitotic G1 phase in cells expressing p53
Inhibits CK1 mediated phosphorylation of LRRK2 in cell lines
Specifications
Chemical Name or Material
1,3-Dihydro-3-[(2,4,6-trimethoxyphenyl)methylene]-2H-indol-2-one
CAS
186611-52-9
Recommended Storage
Store at -20°C
Quantity
5 mg
Molecular Formula
C
18
H
17
NO
4
Formula Weight
311.33
Purity
98%