Polycarbonate Face Shield
Polycarbonate Face Shield
Actual product may vary
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Wear this adjustable polycarbonate face shield over safety glasses. It provides impact protection and features an optically clear 1/16 in. thick polycarbonate window.
Have Questions?
Catalog NumberLens TintLength (English)Width (English)
6355-0001Clear11.6 in.7.8 in.
Catalog number 6355-0001
Price (MYR)
2,461.00
CS
Add to cart
Lens Tint:
Clear
Length (English):
11.6 in.
Width (English):
7.8 in.
Price (MYR)
2,461.00
CS
Add to cart
Wear the adjustable Thermo Scientific™ Nalgene™ Polycarbonate Faceshield over safety glasses. With an optically clear window made of 1/16 in. (1.5mm) thick polycarbonate, this faceshield provides impact protection as well as protection from UV light in the 200-360nm range. It is useful in germicidal procedures as well as reading of chromatographic/electrophoretic strips and gels. Complies with OSHA 29CFR1910.1030 and ANSI Z87.1-1989 standards for use with bloodborne pathogens.
Features:
  • Wraparound design offers both front and side protection while height provides coverage from the bottom of the neck to the top of the head
  • Features a comfortable and adjustable polyethylene headdress
  • Biohazard/Transparent
Note:UV protection from 200 to 360nm (UV radiation may cause the polycarbonate to develop a slight yellow/brown tint.)
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Specifications
Thickness (English)0.060 in.
Thickness (Metric)1.5 mm
Width (English)7.8 in.
DescriptionAdjustable Face Shield
Lens TintClear
Product TypeShield Face PC/PE
Length (English)11.6 in.
No. per Case4/Cs.
MaterialPolycarbonate
Unit SizeCS