Class A PMP Tall-Form Measuring Cylinder
Class A PMP Tall-Form Measuring Cylinder
Actual product may vary
Class A PMP Tall-Form Measuring Cylinder
Manufactured with crystal clear PMP for optimum visibility. VITLab™ Class A PMP Tall-Form Measuring Cylinders are designed with a hexagonal base with bottom studs to provide high stability.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
6.52041000 mL10 mL
Catalog number 6.5204
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
10 mL
  • Raised scale and ring marks at the primary scale points, calibrated ‘In’
  • Lot certificate supplied bears batch number and actual nominal value ascertained under test conditions
  • The resulting deviations from the nominal value fall well under the allowed tolerances of Class A according to DIN 12681 and ISO 6706
  • DE-M marked
  • Laser engraved batch number and year of manufacture
  • Thermal stress up to 121 °C (autoclaving) does not cause tolerance limits to be permanently exceeded
Specifications
Capacity (Metric)1000 mL
MaterialPMP
Tolerance±5 mL
Certifications/ComplianceDIN 12681, ISO 6706
ClassA
ScaleRaised
Diameter (Metric)69 mm
Quantity1Each
Graduations10 mL
AutoclavableAutoclavable
Limit of Error±5mL
ShapeRound
Base TypeHexagonal
Height (Metric)440 mm
TypeGraduated Cylinder
Unit SizeEach