Nalgene™ Unwire™ Polypropylene Test Tube Racks
Nalgene™ Unwire™ Polypropylene Test Tube Racks
Actual product may vary
Nalgene™ Unwire™ Polypropylene Test Tube Racks
Nalgene™ Unwire™ Polypropylene Test Tube Racks
Nalgene™ Unwire™ Polypropylene Test Tube Racks
Nalgene™ Unwire™ Polypropylene Test Tube Racks
Hold a full load of test tubes or centrifuge tubes with these polypropylene test tube racks, molded in one piece.
Have Questions?
Catalog NumberColorFor Use With (Equipment)
5976-0316Blue16mm Tubes
Catalog number 5976-0316
HSN 39269099
Price (MYR)
872.00
Case
Add to cart
Color:
Blue
For Use With (Equipment):
16mm Tubes
Price (MYR)
872.00
Case
Add to cart
Hold a full load of test tubes or centrifuge tubes with Thermo Scientific™ Nalgene™ Unwire™ Polypropylene Test Tube Racks. Racks are molded in one piece to provide extra strength and durability for room temperature applications.
Features:
  • Higher specific gravity than water so rack stays submerged in a water bath
  • Holds a full load of test tubes or centrifuge tubes for 13 and 16 sizes or up to 72 smaller diameter tubes in a 6x12 array
  • Available in four bright colors to help keep samples organized
  • Accepts large labels for easy sample identification
Note:
These racks are not autoclavable.
Nalgene reusable polypropylene(PP) products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These racks and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged. And at end of product life, please wash and recycle your Nalgene racks if services are available in your community.
Specifications
DescriptionTest Tube Rack Unwire™ Blue PP 16mm
AutoclavableNot Autoclavable
ColorBlue
HoldsTubes (16 mm)
MaterialPolypropylene
Positions72
ShapeRectangular
For Use With (Equipment)16mm Tubes
Dimensions (L x W x H)9.75 x 5 x 2.75 in. (24.9 x 12.7 x 6.7 cm)
Length (Metric)24.9 cm
Length (English)9.75 in.
Width (English)5 in.
Height (English)2.75 in.
Diameter (Metric) Opening16 mm
Product TypeTest Tube Rack
Diameter (Metric) Inner16 mm
Shelves3 Tiers
Height (Metric)6.7 cm
Array6 x 12
Width (Metric)12.7 cm
Unit SizeCase
Frequently asked questions (FAQs)
Which Unwire racks are autoclavable?
Any of our ResMer polyketone test tube racks can be autoclaved, by our standard validated parameters (121 degrees Celsius at 15 psig for 20 minutes). Due to their molded structure, our polypropylene Unwire racks (unlike our other polyproylene labware products) cannot be autoclaved.
Which of your Unwire test tube racks can be used for 15 mL or 50 mL sterile conical centrifuge tubes?
Any of our Unwire racks with spaces for tubes 20 mm and 30 mm in diameter can be used, respectively, for 15 mL and 50 mL sterile conical centrifuge tubes.