SUPELCOSIL™ LC-ABZ HPLC Column, 5 μm Particle Size
SUPELCOSIL™ LC-ABZ HPLC Column, 5 μm Particle Size
Actual product may vary
SUPELCOSIL™ LC-ABZ HPLC Column, 5 μm Particle Size
SUPELCOSIL™ LC-ABZ phase has a unique deactivation technology which provides excellent reversed-phase performance for basic compounds, as well as those that are acidic, polar neutral, and non-polar.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
5914225 cm4.6 mm
Catalog number 59142
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
25 cm
Diameter (Metric):
4.6 mm
Silanol suppressing practices such as using competing amines, operating at high ionic strength, and adjusting mobile phases to pH extremes are typically not required to obtain good peak shape on the LC-ABZ phase.
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Specifications
Diameter (Metric)4.6 mm
Length (Metric)25 cm
Particle Size5 μm
Pore Size120 Å
Surface Area170 m2/g
Stationary PhaseAmide, Alkyl
Product LineSUPELCOSIL
MatrixAmide, Alkyl Phase/Silica Gel, Spherical Particle Platform
EndcappedYes
PhaseReversed Phase
Unit SizeBox