Multidrop™ Pico 1 & Pico 8 Digital Dispenser
Multidrop™ Pico 1 & Pico 8 Digital Dispenser
Actual product may vary
Multidrop™ Pico 1 & Pico 8 Digital Dispenser
Miniaturize assays and achieve accurate results on picolter range with non-contact dispensing with eay to use software
Have Questions?
Catalog NumberDescriptionDimensions (L x W x H)
5840600Multidrop Pico 8 Digital Dispenser47 x 38 x 23 cm
Catalog number 5840600
Price (MYR)
-
Description:
Multidrop Pico 8 Digital Dispenser
Dimensions (L x W x H):
47 x 38 x 23 cm
Pico 1 and Pico 8 offers enhanced dispensing precision with volume as low as 11pL in any well, using up to ten times less reagent compared to manual pipetting workflows, enabling significant cost and waste reductions. Designed to enable assay miniaturization, boost productivity and improve accuracy of results makes them ideal for a broad array of low-volume applications such as qPCR, dose response curves, drug screening assays and ELISA.
Improved Efficiency and Speed
The Digital Dispenser offers 40 times greater speed than analog serial dilution – from 10 minutes to just 15 seconds – eliminating precious time and accelerating your research.
Features:
  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software
The Digital Dispenser supports a wide range of applications and sample types:
  • qPCR
  • Drug combination studies
  • DMPK
  • Assay development
  • Biochemical assays
  • Secondary screening
  • Cell-based assays
  • Spotting
  • Enzyme profiling
  • Synthetic biology
  • Antibody therapies
  • SAR
Improved Efficiency and Speed
The Digital Dispenser offers 40 times greater speed than analog serial dilution – from 10 minutes to just 15 seconds – eliminating precious time and accelerating your research.
Specifications
Product TypeDigital Dispenser
DescriptionMultidrop Pico 8 Digital Dispenser
Volume (Metric)11 pL to 20 μL (8-well Cartridge), 1 nL to 200 μL (4-well Cartridge)
For Use With (Application)Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
InterfaceUSB
Plate Type12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
Precision≤ 8%
SoftwarePicoIT 8
Speed11 pL to 10 μL in seconds
Dimensions (L x W x H)47 x 38 x 23 cm
Weight (Metric)14.1
Electrical Requirements100 to 240V 50/60Hz
Voltage100/240 V
Frequency50/60 Hz
Unit SizeEach
Frequently asked questions (FAQs)
Do the Multidrop Pico 1 & Pico 8 Digital Dispensers come with a laptop?
No, Multidrop Pico 1 & Pico 8 Digital Dispensers are not sold with a laptop. This would need to be purchased separately. You can also use your own laptop or PC, as long as it meets the minimum PC requirements outlined in Chapter 3 of the Thermo Scientific Multidrop Pico 1 or Multidrop Pico 8 Operating Manuals: