T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Actual product may vary
T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
T-865 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Perform a variety of applications with the Thermo Scientific™ T-865 Fixed Angle Rotor, which offers eight-place capacity.
Have Questions?
Catalog NumberCapacity
514118 x 36 mL
Catalog number 51411
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
8 x 36 mL
Perform a variety of applications with the Thermo Scientific™ T-865 Fixed Angle Rotor, which offers eight-place capacity.
Recommended for:
Differential protein fractionation, DNA isolation.
Specifications
DescriptionT-865 Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity8 x 36 mL
Max. RCF429,459 x g
Max. Speed65,000 rpm
Radius (Metric)91 mm
Radius (Metric) Min.38.4 mm
K Factor51.7
Max. Tube Length97 mm
Max. Tube Diameter25 mm
Net Weight18.3 lb. (8.3 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle23.5°
Avg. RCF305,340 x g
Min. RCF181,222 x g
Unit SizeEA